(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid 95%

Supplier: Ambeed
Warning

A119584-100G A119584-1G A119584-25G A119584-10G
103799-786EA 10 USD
103799-786 103830-610 77094-412 77311-906 77685-054 77901-848
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid 95%
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid

Caution: for Research Use Only

Formula: C₁₆H₂₂N₂O₆
Boiling Pt: 548.7 °C (760 mm Hg)
Flash Pt: 285.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237342
CAS Number: 16947-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR