7-Methoxyisoflavone ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-3-phenyl-4H-chromen-4-one

A860031-100MG A860031-1G A860031-250MG
77614-542EA 52.53 USD
77614-542 77614-544 77684-820
7-Methoxyisoflavone ≥98%, Ambeed.Inc
7-Methoxyisoflavone

Caution: For research use only.

Formula: C₁₆H₁₂O₃
MDL Number: MFCD00181416
CAS Number: 1621-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR