1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 1,2-Bis(phenylsulphinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethane palladium(II) acetate, 1,2-Bis(phenylsulphinyl)ethane palladium(II) acetate, White Catalyst, Christina White Catalyst

A445804-100MG A445804-250MG A445804-5G
77684-222EA 61 USD
77684-222 77684-223 77886-182
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97%, Ambeed.Inc
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate

Caution: For research use only.

Formula: C₁₆H₁₈O₄PdS₂
MDL Number: MFCD09842752
CAS Number: 858971-43-4
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR