5-Methoxynicotinaldehyde 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A174163-10G
103806-926EA 14 USD
103806-926 103845-224 77683-400
5-Methoxynicotinaldehyde 97%, Ambeed.Inc
5-Methoxynicotinaldehyde

Caution: For research use only.

Formula: C7H7NO2
Melting Pt: 28…32 °C
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07437946
CAS Number: 113118-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR