DAPT 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
A150458-10MG
A150458-1MG
A150458-5MG
A150458-100MG
A150458-1G
A150458-250MG
77162-746EA
41
USD
77162-746
77162-748
77162-750
77683-269
77683-270
77683-271
DAPT 98%, Ambeed.Inc
DAPT
Caution: For research use only.
Formula:
C₂₃H₂₆F₂N₂O₄ |
MDL Number:
MFCD04974585 CAS Number: 208255-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...