Dianilinopyrimidine-01 ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: CZC-8004, N2-(4-(Aminomethyl)phenyl)-5-fluoro-N4-phenylpyrimidine-2,4-diamine

A114366-1G
77683-038EA 566 USD
77683-038
Dianilinopyrimidine-01 ≥95%, Ambeed.Inc
Dianilinopyrimidine-01

Caution: For research use only.

Formula: C₁₇H₁₆FN₅
MDL Number: MFCD16251195
CAS Number: 916603-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR