Gold ≥99.95%, disk, as rolled, Diameter 19.5 mm

Supplier: GOODFELLOW CORPORATION

1000044050 1000046750 1000046959 1000043559
77681-376EA 835.22 USD
77681-376 77681-738 77681-812 77681-301
Gold ≥99.95%, disk, as rolled, Diameter 19.5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR