Gold ≥99.95%, disk, as rolled, Diameter 13 mm

Supplier: GOODFELLOW CORPORATION

1000044014 1000043532 1000046708 1000046917
77681-362EA 491.43 USD
77681-362 77681-287 77681-724 77681-798
Gold ≥99.95%, disk, as rolled, Diameter 13 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR