Gold ≥99.95%, disk, as rolled, Diameter 44 mm
Supplier: GOODFELLOW CORPORATION
1000044123
1000043626
1000046843
1000047066
77681-408EA
2763.28
USD
77681-408
77681-335
77681-772
77681-847
Gold ≥99.95%, disk, as rolled, Diameter 44 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...