Gold ≥99.95%, disk, as rolled, Diameter 24.8 mm

Supplier: GOODFELLOW CORPORATION

1000044074 1000043578 1000046790 1000046993
77681-388EA 1173.15 USD
77681-388 77681-312 77681-750 77681-824
Gold ≥99.95%, disk, as rolled, Diameter 24.8 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR