Gold ≥99.95%, disk, as rolled, Diameter 18 mm

Supplier: GOODFELLOW CORPORATION

1000044039 1000046738 1000043551 1000046949
77681-372EA 748.64 USD
77681-372 77681-734 77681-297 77681-808
Gold ≥99.95%, disk, as rolled, Diameter 18 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR