Gold ≥99.95%, disk, as rolled, Diameter 17.4 mm
Supplier: GOODFELLOW CORPORATION
1000044037
1000046734
1000043549
1000046945
77681-371EA
715.18
USD
77681-371
77681-733
77681-296
77681-807
Gold ≥99.95%, disk, as rolled, Diameter 17.4 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...