Gold ≥99.99%, disk, as rolled, Diameter 6.5 mm

Supplier: GOODFELLOW CORPORATION

1000046452 1000047396
77681-633EA 298.44 USD
77681-633 77681-856
Gold ≥99.99%, disk, as rolled, Diameter 6.5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR