Gold ≥99.999%, disk, as rolled, Diameter 40 mm

Supplier: GOODFELLOW CORPORATION

1000031134 1000031876 1000031233 1000044985
77680-963EA 3804.4 USD
77680-963 77681-119 77680-986 77681-552
Gold ≥99.999%, disk, as rolled, Diameter 40 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR