Gold ≥99.999%, disk, as rolled, Diameter 20.7 mm

Supplier: GOODFELLOW CORPORATION

1000041449 1000044236 1000044931 1000046332
77681-246EA 5256.46 USD
77681-246 77681-453 77681-525 77681-594
Gold ≥99.999%, disk, as rolled, Diameter 20.7 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR