Gold ≥99.999%, disk, as rolled, Diameter 12.1 mm
Supplier: GOODFELLOW CORPORATION
1000046280
1000041401
1000044194
1000044892
77681-578EA
477.59
USD
77681-578
77681-227
77681-434
77681-506
Gold ≥99.999%, disk, as rolled, Diameter 12.1 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...