Gold ≥99.999%, disk, as rolled, Diameter 10.5 mm

Supplier: GOODFELLOW CORPORATION

1000046268 1000041389 1000044882 1000037927
77681-572EA 403.96 USD
77681-572 77681-221 77681-501 77681-200
Gold ≥99.999%, disk, as rolled, Diameter 10.5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR