Gold ≥99.999%, disk, as rolled, Diameter 5 mm

Supplier: GOODFELLOW CORPORATION

1000029071 1000044152 1000041358 1000044851
77680-731EA 203.11 USD
77680-731 77681-414 77681-206 77681-488
Gold ≥99.999%, disk, as rolled, Diameter 5 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR