Gold ≥99.95%, disk, as rolled, Diameter 50 mm
Supplier: GOODFELLOW CORPORATION
1000031083
1000030893
1000031503
1000032003
1000030634
1000030757
1000031956
1000032112
1000044758
77680-951EA
3899.65
USD
77680-951
77680-911
77681-034
77681-163
77680-839
77680-881
77681-149
77681-192
77681-484
Gold ≥99.95%, disk, as rolled, Diameter 50 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...