Gold ≥99.95%, disk, as rolled, Diameter 21 mm
Supplier: GOODFELLOW CORPORATION
1000046973
1000043565
1000044059
1000046770
77681-817EA
2384.85
USD
77681-817
77681-305
77681-381
77681-743
Gold ≥99.95%, disk, as rolled, Diameter 21 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...