Gold ≥99.95%, disk, as rolled, Diameter 14.5 mm
Supplier: GOODFELLOW CORPORATION
1000043542
1000044027
1000046933
1000046723
77681-292EA
384.44
USD
77681-292
77681-367
77681-803
77681-729
Gold ≥99.95%, disk, as rolled, Diameter 14.5 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...