Gold ≥99.95%, disk, as rolled, Diameter 12 mm

Supplier: GOODFELLOW CORPORATION

1000017221 1000043118 1000043526 1000046700 1000046907
77680-711EA 446.17 USD
77680-711 77681-262 77681-285 77681-722 77681-796
Gold ≥99.95%, disk, as rolled, Diameter 12 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR