Gold ≥99.999%, disk, as rolled, Diameter 19 mm

Supplier: GOODFELLOW CORPORATION

1000041437 1000044225 1000044921 1000046318
77681-241EA 4604.35 USD
77681-241 77681-448 77681-520 77681-590
Gold ≥99.999%, disk, as rolled, Diameter 19 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR