Gold ≥99.999%, disk, as rolled, Diameter 18.8 mm
Supplier: GOODFELLOW CORPORATION
1000041435
1000044920
1000044223
1000046314
77681-240EA
4529.89
USD
77681-240
77681-519
77681-447
77681-589
Gold ≥99.999%, disk, as rolled, Diameter 18.8 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...