Gold ≥99.999%, disk, as rolled, Diameter 13.7 mm

Supplier: GOODFELLOW CORPORATION

1000044202 1000041410 1000044898 1000046288
77681-437EA 779.31 USD
77681-437 77681-230 77681-509 77681-580
Gold ≥99.999%, disk, as rolled, Diameter 13.7 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR