Gold ≥99.99%, disk, as rolled, Diameter 4 mm
Supplier: GOODFELLOW CORPORATION
1000031918
1000003569
1000013264
1000030564
1000031577
1000032081
77681-134EA
424.32
USD
77681-134
77680-693
77680-702
77680-822
77681-046
77681-176
Gold ≥99.99%, disk, as rolled, Diameter 4 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...