Gold ≥99.95%, disk, as rolled, Diameter 6.35 mm

Supplier: GOODFELLOW CORPORATION

1000031085 1000031958 1000030896 1000031506 1000032004 1000030635 1000030759 1000031289
77680-952EA 234.56 USD
77680-952 77681-150 77680-912 77681-035 77681-164 77680-840 77680-882 77681-002
Gold ≥99.95%, disk, as rolled, Diameter 6.35 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR