Gold ≥99.95%, disk, as rolled, Diameter 40 mm

Supplier: GOODFELLOW CORPORATION

1000030749 1000030884 1000031076 1000031495 1000031997 1000032107 1000030629 1000031284 1000031950
77680-878EA 1607.04 USD
77680-878 77680-908 77680-948 77681-032 77681-160 77681-189 77680-836 77681-000 77681-146
Gold ≥99.95%, disk, as rolled, Diameter 40 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR