Gold ≥99.95%, disk, as rolled, Diameter 50 mm

Supplier: GOODFELLOW CORPORATION

1000031083 1000030893 1000031503 1000032003 1000030634 1000030757 1000031956 1000032112 1000044758
77680-951EA 3899.65 USD
77680-951 77680-911 77681-034 77681-163 77680-839 77680-881 77681-149 77681-192 77681-484
Gold ≥99.95%, disk, as rolled, Diameter 50 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR