Gold ≥99.95%, disk, as rolled, Diameter 4 mm

Supplier: GOODFELLOW CORPORATION

1000030752 1000030887 1000001033 1000031078 1000031999 1000030630 1000031286 1000031707 1000031951 1000032109
77680-879EA 228.55 USD
77680-879 77680-909 77680-690 77680-949 77681-161 77680-837 77681-001 77681-069 77681-147 77681-190
Gold ≥99.95%, disk, as rolled, Diameter 4 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR