Gold ≥99.9%, disk, Diameter 4 mm

Supplier: GOODFELLOW CORPORATION

1000030319 1000030420 1000030373 1000030976 1000030526 1000031023 1000030937 1000031180 1000031746 1000030470 1000031831 1000030817 1000031332 1000031637 1000031786
77680-741EA 128.33 USD
77680-741 77680-782 77680-767 77680-926 77680-809 77680-935 77680-918 77680-974 77681-080 77680-797 77681-105 77680-894 77681-009 77681-060 77681-091
Gold ≥99.9%, disk, Diameter 4 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR