Gold ≥99.9%, disk, Diameter 15 mm

Supplier: GOODFELLOW CORPORATION

1000030407 1000030364 1000030973 1000012845 1000030516 1000031013 1000030930 1000031177 1000030308 1000030460 1000030799 1000031620 1000031734 1000031329 1000031821
77680-776EA 315.82 USD
77680-776 77680-761 77680-925 77680-701 77680-803 77680-933 77680-917 77680-973 77680-735 77680-791 77680-888 77681-055 77681-075 77681-008 77681-099
Gold ≥99.9%, disk, Diameter 15 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR