Gold ≥99.9%, disk, Diameter 25.4 mm

Supplier: GOODFELLOW CORPORATION

1000030411 1000030368 1000030519 1000031780 1000030312 1000030463 1000031626 1000031738 1000031825 1000030805
77680-778EA 403.83 USD
77680-778 77680-763 77680-805 77681-088 77680-737 77680-793 77681-057 77681-077 77681-101 77680-890
Gold ≥99.9%, disk, Diameter 25.4 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR