Gold ≥99.99%, disk, as rolled, Diameter 8 mm

Supplier: GOODFELLOW CORPORATION

1000030675 1000031922 1000030706 1000032089 1000030571 1000031590
77680-856EA 279.23 USD
77680-856 77681-136 77680-868 77681-181 77680-827 77681-051
Gold ≥99.99%, disk, as rolled, Diameter 8 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR