Gold ≥99.99%, disk, as rolled, Diameter 30 mm

Supplier: GOODFELLOW CORPORATION

1000030664 1000031915 1000030696 1000030561 1000031571 1000032078
77680-850EA 617.97 USD
77680-850 77681-132 77680-863 77680-820 77681-044 77681-174
Gold ≥99.99%, disk, as rolled, Diameter 30 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR