Gold ≥99.99%, disk, as rolled, Diameter 25.4 mm

Supplier: GOODFELLOW CORPORATION

1000030559 1000031912 1000030693 1000030661 1000031565 1000032075
77680-819EA 392.81 USD
77680-819 77681-130 77680-861 77680-848 77681-042 77681-172
Gold ≥99.99%, disk, as rolled, Diameter 25.4 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR