Gold ≥99.99%, disk, as rolled, Diameter 12.5 mm
Supplier: GOODFELLOW CORPORATION
1000030552
1000030687
1000031554
1000032068
1000030655
1000031907
77680-814EA
204.85
USD
77680-814
77680-858
77681-038
77681-168
77680-844
77681-127
Gold ≥99.99%, disk, as rolled, Diameter 12.5 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...