Gold ≥99.95%, disk, as rolled, Diameter 15 mm

Supplier: GOODFELLOW CORPORATION

1000030731 1000030869 1000031059 1000031991 1000031273 1000032101 1000030620 1000031481 1000031701 1000031940
77680-872EA 522.6 USD
77680-872 77680-903 77680-942 77681-156 77680-995 77681-185 77680-831 77681-028 77681-067 77681-140
Gold ≥99.95%, disk, as rolled, Diameter 15 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR