Gold ≥99.95%, disk, as rolled, Diameter 12.7 mm

Supplier: GOODFELLOW CORPORATION

1000030728 1000030866 1000031939 1000031056 1000031989 1000031270 1000032099 1000030618 1000031478
77680-871EA 312.06 USD
77680-871 77680-902 77681-139 77680-941 77681-155 77680-994 77681-184 77680-830 77681-027
Gold ≥99.95%, disk, as rolled, Diameter 12.7 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR