Gold ≥99.999%, disk, as rolled, Diameter 20 mm

Supplier: GOODFELLOW CORPORATION

1000030339 1000031122 1000031387 1000031869 1000031224
77680-751EA 4983.64 USD
77680-751 77680-959 77681-018 77681-115 77680-982
Gold ≥99.999%, disk, as rolled, Diameter 20 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR