Gold ≥99.95%, disk, as rolled, Diameter 30 mm

Supplier: GOODFELLOW CORPORATION

1000030743 1000030881 1000031071 1000031996 1000031282 1000031492 1000032106 1000030625 1000031947
77680-876EA 1114.21 USD
77680-876 77680-907 77680-946 77681-159 77680-999 77681-031 77681-188 77680-834 77681-144
Gold ≥99.95%, disk, as rolled, Diameter 30 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR