Gold ≥99.95%, disk, as rolled, Diameter 25.4 mm

Supplier: GOODFELLOW CORPORATION

1000030737 1000030875 1000016278 1000031065 1000031993 1000031278 1000031487 1000031944
77680-874EA 712.5 USD
77680-874 77680-905 77680-710 77680-944 77681-157 77680-997 77681-030 77681-142
Gold ≥99.95%, disk, as rolled, Diameter 25.4 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR