Gold ≥99.999%, disk, as rolled, Diameter 8 mm

Supplier: GOODFELLOW CORPORATION

1000031152 1000030350 1000031248 1000031886 1000031404
77680-969EA 395.87 USD
77680-969 77680-757 77680-992 77681-125 77681-025
Gold ≥99.999%, disk, as rolled, Diameter 8 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR