Gold ≥99.9%, disk, Diameter 12.7 mm

Supplier: GOODFELLOW CORPORATION

1000030405 1000030362 1000030970 1000030514 1000031010 1000030927 1000031174 1000031775 1000030306 1000030458 1000030796 1000031617 1000031733 1000031326 1000031820
77680-775EA 215.66 USD
77680-775 77680-760 77680-924 77680-802 77680-932 77680-916 77680-972 77681-086 77680-734 77680-790 77680-887 77681-054 77681-074 77681-007 77681-098
Gold ≥99.9%, disk, Diameter 12.7 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR