Gold ≥99.95%, disk, as rolled, Diameter 6 mm

Supplier: GOODFELLOW CORPORATION

1000031088 1000031961 1000030899 1000031509 1000032006 1000030639 1000030761 1000031710 1000031292 1000032114
77680-953EA 403.12 USD
77680-953 77681-151 77680-913 77681-036 77681-165 77680-841 77680-883 77681-070 77681-003 77681-193
Gold ≥99.95%, disk, as rolled, Diameter 6 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR