Gold ≥99.95%, disk, as rolled, Diameter 25.4 mm
Supplier: GOODFELLOW CORPORATION
1000030737
1000030875
1000016278
1000031065
1000031993
1000031278
1000031487
1000031944
77680-874EA
712.5
USD
77680-874
77680-905
77680-710
77680-944
77681-157
77680-997
77681-030
77681-142
Gold ≥99.95%, disk, as rolled, Diameter 25.4 mm
Gold
Formula:
Au Boiling Pt: 3080 °C Melting Pt: 1064.4 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...