Gold ≥99.95%, disk, as rolled, Diameter 12.5 mm

Supplier: GOODFELLOW CORPORATION

1000030725 1000030863 1000031937 1000031053 1000031987 1000031267 1000032097 1000030616 1000031475
77680-870EA 307.71 USD
77680-870 77680-901 77681-138 77680-940 77681-154 77680-993 77681-183 77680-829 77681-026
Gold ≥99.95%, disk, as rolled, Diameter 12.5 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR