Gold ≥99.99%, disk, as rolled, Diameter 25 mm

Supplier: GOODFELLOW CORPORATION

1000031914 1000007256 1000030694 1000030662 1000031568 1000032077
77681-131EA 2235.23 USD
77681-131 77680-698 77680-862 77680-849 77681-043 77681-173
Gold ≥99.99%, disk, as rolled, Diameter 25 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR