Gold ≥99.99%, disk, as rolled, Diameter 20 mm

Supplier: GOODFELLOW CORPORATION

1000030557 1000030692 1000032073 1000030659 1000031562 1000031911
77680-818EA 304.49 USD
77680-818 77680-860 77681-171 77680-847 77681-041 77681-129
Gold ≥99.99%, disk, as rolled, Diameter 20 mm
Gold
Formula: Au
Boiling Pt: 3080 °C
Melting Pt: 1064.4 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR