Gold ≥99.99%, disk, as rolled, Diameter 12.5 mm

Supplier: GOODFELLOW CORPORATION

1000030552 1000030687 1000031554 1000032068 1000030655 1000031907
77680-814EA 204.85 USD
77680-814 77680-858 77681-038 77681-168 77680-844 77681-127
Gold ≥99.99%, disk, as rolled, Diameter 12.5 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR